Learn More
Sperian™ Honeywell™ Laser Lite LS-400 Earplug Refill
Makes protection fun and highly visible. Sperian™ Honeywell™ Laser Lite LS-400 Earplug Refills are made from self-adjusting polyurethane foam which fits virtually every wearer.
Brand:  Sperian™ 1013047_CLEAN
Product Code. 12350160
  • £64.05 / 200 pairs
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Vibrant colors
  • Contoured t-shape aids in easy insertion and removal from ear canal
  • Comfortable long-term wear
  • Ideal for process industries with low tolerances for packaging waste
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yellow, Magenta
200/Pk.
Earplug
Universal
T-shaped