Learn More
Sperian™ Honeywell™ Quiet Uncorded Earplugs
Click to view available options
Color: Orange
Description
- No-roll design
- Easy to handle and fit into ear canal
- Built-in insertion stem
- Smooth and non-irritating to skin
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Orange |
Size | Regular |
Quantity | 50/Pk. |
Plug Shape | Bell |
Product Type | Ear Plugs |