Learn More
Brady™ Honeywell Enhanced Xenon 1950H HD Handheld Healthcare Scanner
The Xenon 1950H HD scanner delivers superior barcode scanning and digital image capture.
Brand: Brady™ 306801
522.50 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- The scanner incorporates a revolutionary decoding architecture and a custom sensor, enabling extended depth of field, faster reading and improved scanning performance on poor-quality barcodes
- From high density linear to 2D barcodes found directly on the screen of a mobile device, the 1950H HD scanner decodes virtually all barcodes with ease
Specifications
High Density Imager Sensor | |
USB | |
500 mA (at 5 V DC) | |
XENON 1950HHD |