Learn More
Sperian™ Honeywell™ ARMAMAX AX™ Clear Lens Safety Goggles
Product Code. 11847221
Click to view available options
Lens Coating:
Hardcoat
Uncoated
Unit Size:
Each
Pack of 10
Product Code. 11847221
Brand: Sperian™ 1002221
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Offering high levels of comfort and support. Sperian™ Honeywell™ ARMAMAX AX™ Clear Lens Safety Goggles fit comfortably over prescription eyewear.
  • Practical and durable
  • Frosted side panels and browguard sections reduce glare/reflections
  • Curved temples and a raised nose bridge offer a precise fit
  • Available with or without hard lens coating
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001 / 2000, EN 166 1F
Lens Coating Hardcoat
Lens Tint Clear
Product Type Eye and Face Protection Spectacles
For Use With (Application) Over-the-glass spectacle, General use and protection against impact (45m/s), Mechanic work: Grinding, turning, milling
Frame Color Transparent
No. per Pack 10/Pk.
Lens Material Polycarbonate