missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Freedom Half-Mask Air-Purifying Respirator (APR)
The Freedom Half-Mask APR is designed for occasional exposure to gases, vapours, and particulates. Limited use and maintenance-free, it is supplied pre-assembled with cartridges for elimination of assembly mistakes, replacement parts, and decontamination concerns.
£16.43 - £29.74
Specifications
Design Type | Cup/Molded |
---|---|
Disposable | Reusable |
For Use With (Equipment) | Prefilter Pads [11847171 (P1), 12682743 (P2)] and Pad Retaining Covers (12662653) |
Strap Attachment Type | One-Piece Sliding Suspension System |
Alert:
To be disposed of when filters are saturated.
Product Code | Brand | Cartridge/Filter | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cartridge/Filter | Price | Quantity & Availability | |||||
11887151
|
Honeywell™
1001591 |
A1 | N/A | ||||||
11887231
|
Honeywell™
1001598 |
A2 | N/A | ||||||
11897151
|
Honeywell™
1001592 |
A1P1 | N/A | ||||||
11817161
|
Honeywell™
1001597 |
P3 | N/A | ||||||
11807161
|
Honeywell™
1001602 |
A2P3 | N/A | ||||||
11817010
|
Honeywell™
1001610 |
A1B1E1K1P3 | N/A | ||||||
Description
- Product Type: Air-Purifying Respirator (APR)
- Mask Type: Half Mask
- Standards: EN405
- Cartridge Type: Dual Cartridge, Pre-Assembled
- Fastenings: One-Piece Sliding Suspension System
- Hazards: Gases, Organic Vapors (Temp. >65°C), Inorganic Vapors, Acids, Ammoniac and Derived Aminates (Class 1); Solid and Liquid Particulates (Class 3)
- For Use With (Equipment): Prefilter Pads (11847171, 12682743) and Pad Retaining Covers (12662653)
- For Use With (Applications): Chemical Industry, Construction Industry, Food Services, Manufacturing Industry, Medical Industry, Pharmaceutical Industry
Specifications
Cup/Molded | |
Reusable | |
Prefilter Pads [11847171 (P1), 12682743 (P2)] and Pad Retaining Covers (12662653) | |
One-Piece Sliding Suspension System |