Learn More
Honeywell™ Supervizor Face Shields
Robust, heavy-duty, helmet-mounted face shield range with hearing protection compatibility. Sperian™ Honeywell™ Supervizor Face Shields have a wide visor selection that can be tailored to the specific hazard/application.
Brand:  Honeywell™ 1002312
 View more versions of this product
Product Code. 11826990
Quantity Price
1 £24.32 / Each
10 £20.60 / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Recommended for use in the chemical and forestry industries, among others
  • Protects against hazards such as abrasion, airborne particles, biohazards, chemicals, gases, vapors, and smoke
  • Maximum protection, allowing full unrestricted head movement
  • Creased visor eliminates gaps and visor profile follows contours of the face
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN 166 1B, EC 0100, EC 0086
230 mm Clear Polycarbonate Visor
SV9PC
Polycarbonate
230 mm
Supervizor Faceshield
1 Ea.