Learn More
Honeywell™ Optifit Full Face Mask
Effective respiratory protection in a full-face mask. Honeywell™ Optifit Full Face Mask offers dedicated respiratory protection depending on the canister type assembled to it.
Brand:  Honeywell™ 1715011
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11896950
  • £282.00 / Each
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Reusable APR with respiratory protection dictated by canister type selected.
  • For optimal protection, Sperian™ Aluminum Canisters or Sperian™ Plastic Canisters are recommended.
  • Note: Can only be used in sufficiently ventilated premises where the atmoshpere contains a minimum oxygen volume of 17%.
  • U-shaped silicone skirt.
  • Full lens with optimal lateral and downward vision.
  • Free choice of filter
  • Easy care
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Filter
Optifit Full Face Mask
Optifit
Silicone Skirt/Dual Filter/ Diaphragm
EC Category PPE: 3, Quality Assurance: ISO 9001/2000, EC Certification Number: 0070-003-079-01-02-0009 - Ext. 01-10-03, EC Attestation Number 0070-003-079-01-02-0009 - Ext. 01-10-03, Declaration of ocnformity: INRS-1715011, EN 136
Medium
For industry uses
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Honeywell™ Optifit Full Face Mask > Size: Medium