Learn More
Nabertherm™ Horizontal High-Temperature Tube Furnace
High-temperature horizontal tube furnaces. High-quality insulation materials made of vacuum-formed fiber plates enable energy-saving operation and a fast heating time due to low heat storage and heat conductivity. Different gas supply systems available.
£15574.82 - £29015.64
Product Code | Brand | Diameter (Metric) Process Tube | Temperature (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Process Tube | Temperature (Metric) | Price | Quantity & Availability | ||||
17308119
|
Nabertherm™
RH282V82AN |
80 mm | Up to 1800°C |
£22639.42
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
17358109
|
Nabertherm™
RH152V62AN |
50 mm | Up to 1600°C |
£15574.82
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
17368109
|
Nabertherm™
RH152V72AN |
50 mm | Up to 1700°C |
£17476.50
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
17378109
|
Nabertherm™
RH152V82AN |
50 mm | Up to 1800°C |
£19111.43
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
17388109
|
Nabertherm™
RH282V62AN |
80 mm | Up to 1600°C |
£17510.92
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
17318119
|
Nabertherm™
RH322V62AN |
120 mm | Up to 1600°C |
£22123.13
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
17328119
|
Nabertherm™
RH322V72AN |
120 mm | Up to 1700°C |
£25126.23
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
17338119
|
Nabertherm™
RH322V82AN |
120 mm | Up to 1800°C |
£29015.64
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
17398109
|
Nabertherm™
RH282V72AN |
80 mm | Up to 1700°C |
£20290.29
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
Beschreibung
- Use: horizontal
- Electrical connection: 3-phase
Spezifikation
P570 | |
Horizontal |