Learn More
Scientific Industries Horizontal Orbital Shaker Attachment for Enviro-Genie and Incubator-Genie
An accessory that adds horizontal orbital shaking capability to the Enviro-Genie and the Incubator-Genie.
Brand: Scientific Industries SI-1275
Additional Details : Weight : 1.80000kg
Description
- Adds horizontal orbital shaking capability to the Enviro-Genie and the Incubator-Genie with variable speed of 15-100 rpm
- Ideal for Western Blots and for maintaining cells or solutions in suspension
- No tools required
- Simply remove the Magnetic Platform from the Enviro-Genie or Incubator-Genie and attach the Horizontal Orbital Shaker Attachment
- Fits all new and existing Enviro and Incubator models
Specifications
2 kg | |
13 in. | |
13 x 13 in. | |
5 in. | |
13 in. | |
13 in. | |
3.5 lb. | |
Black | |
Orbital Shaking of Various Containers Including Flasks and Microplates. | |
Horizontal Orbital Shaker Attachment with Texture Mat | |
1 Ea. |
Various containers including flasks and microplates | |
330.20 mm | |
13 x 13 x 5 in. | |
127 mm | |
330.20 mm | |
330.20 mm | |
1.6 kg | |
Horizontal Orbital Shaker for Enviro-Genie and Incubator-Genie | |
Scientific Industries Enviro-Genie and Incubator-Genie | |
1 |