Learn More
Scientific Industries SI™ Horizontal Tube Holder
Product Code. 15527355
Click to view available options
Holds:
12 x 15 mL Tubes
24 Microtubes
50 mL Tubes
Unit Size:
Each
Product Code. 15527355
Brand: Scientific Industries SI™ 83041274
Only null left
Add to basket
Add to basket
Snap tubes into place swiftly and easily. Scientific Instruments SI™ Horizontal Tube Holder does not require additional attachments for use.
  • Holds tubes in place horizontally.
  • Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
  • No additional attachments are necessary
  • Holds tubes in place horizontally.
  • Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
  • No additional attachments are necessary
Specifications
Holds 24 Microtubes
For Use With (Equipment) Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
No. of Positions 24