missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Hormones: 3,3',5-Triiodo-L-Thyronine
£52.95
Specifications
Form | Crystalline powder |
---|---|
Product Type | Progesterone |
Sterility | Sterile |
pH | 9 |
Source | Human |
Description
For research use only. Not intended for therapeutic use.
Specifications
Crystalline powder | |
Sterile | |
Human |
Progesterone | |
9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Hormones: 3,3',5-Triiodo-L-Thyronine