missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Hormones: b-Estradiol

Product Code. p-4882229 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1000 mg
5000 mg
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11424942

Brand: MP Biomedicals™ 0219456505

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

For research use only. Not intended for therapeutic use.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Form Crystalline powder
Product Type Beta-Estradiol
Sterility Sterile
Quantity 5000 mg
compliance-icons
Product Identifier
  • beta-Estradiol, cell culture reagent
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H362-May cause harm to breast-fed children.
  • H373-May cause damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P263-Avoid contact during pregnancy/while nursing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : estradiol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.