missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Horse anti-Mouse M.O.M.(R) (Mouse-On-Mouse) ImmPRESS(R) IgG Secondary Antibody, HRP Polymer, Novus Biologicals™
Horse Polyclonal Antibody
Brand: Novus Biologicals MPX-2402-NB
187.20 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
M.O.M.(R) (Mouse-On-Mouse) ImmPRESS(R) IgG Polyclonal antibody specifically detects M.O.M.(R) (Mouse-On-Mouse) ImmPRESS(R) IgG in Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry
Specifications
M.O.M.(R) (Mouse-On-Mouse) ImmPRESS(R) IgG | |
Polyclonal | |
Equine | |
Affinity purified | |
RUO | |
Mouse | |
Purified |
Immunohistochemistry, Immunocytochemistry | |
HRP Polymer | |
Mouse IgG | |
15 mL | |
Secondary | |
Store at 4°C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Horse anti-Mouse M.O.M.(R) (Mouse-On-Mouse) ImmPRESS(R) IgG Secondary Antibody, HRP Polymer, Novus Biologicals™ > 15 mL; HRP Polymer
Spot an opportunity for improvement?Share a Content Correction