missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hot Gas Bypass Temperature Controllers

Combine these hot gas bypass temperature controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.

Brand:  Thermo Scientific™ 195700

 View more versions of this product

Product Code. 11674210

  • £2950.00 / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combine Thermo Scientific™ Hot Gas Bypass Temperature Controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.

Features:
  • 120V or 220V
  • Factory or field installed

Ordering Information:

Field installed option must be installed by a qualified professional.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Field installed (requires qualified professional); 120V
Field-Installed
CryoPlus 1,2,3,4
120/220 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Hot Gas Bypass Temperature Controllers >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.