missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Red Hot Grip

Kartell™ Red Hot Grib provides protection when working with hot and cold temperatures.

Brand:  Kartell™ 0391500

Product Code. 10216531

  • £31.30 / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For protection against hot containers up to 260°C
  • For protection against cold containers down to -57°C
  • Internal relief surface
Specifications

Specifications

Red
Silicon
Hot and cold handling
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers