missing translation for 'onlineSavingsMsg'
Learn More

Gestigkeit Harry Hotplate with Performance Control

Hotplate with Performance Control are robust and highly durable in non-stop operation.

Brand:  Gestigkeit Harry HD2

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10529131

  • £1420.00 / Each
Estimated Shipment: 10-10-2024
to see stock.
Add to basket

Description

Description

  • Electrical hotplates with variable temperature control, with or without a thermostatic controller. Robust construction for continuous perfomance. aluminum alloy hotplate.
  • Because of the asymmetrical, long-term heating system (hotplate corners and edges are subjected to more heat) an even temperature is guaranteed over the entire heating surface.
  • Asbest-free insulation. The housing is constructed from high-grade, stainless steel No. 1.4301, whereby the central section has been additionally sprayed in orange.
  • Four adjustable feet ensure extra stability. Connecting cable approx. 1,7 m. long, 230 Volt/ 3200 Watt with Schuko plug.
Specifications

Specifications

Hotplate
230 V
Aluminum Alloy
170 mm
18 kg
2850 W
364 x 514 x 170 mm
Stainless Steel
Up to 370°C
364 mm
514 mm
Performance Control
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers