Learn More
Fisherbrand™ Support Rod
For Use With: Isotemp hotplates, stirrers and stirring hotplates
Brand: Fisherbrand™ CIC0001832
30.06 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Fisherbrand Hotplate/Stirrer Support Rod is to be used together with clamps and clamp holder to hold apparatus suitable for Isotemp hotplates, stirrers and stirring hotplates
- Support rod
Specifications
Isotemp hotplates, stirrers and stirring hotplates | |
Use together with clamps and clamp holder to hold apparatus | |
Rod |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30