Learn More
Thermo Scientific™ HotStart™ Storage Reaction Tubes
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
£270.14 - £836.40
Specifications
For Use With (Equipment) Thermal Cyclers
Sterility Sterile
View More Specs
Products 2
Product Code Brand Description Volume (Metric) Price Quantity & Availability  
Product Code Brand Description Volume (Metric) Price Quantity & Availability  
15292559
View Documents
Thermo Scientific™
6210
HotStart Micro 50 Strips 0.2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12318222
View Documents
Thermo Scientific™
6310-11
Hot Start Micro 50 Bulk 0.2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Features:
  • Bead is stable at room temperature
  • Assures synchronous reaction startup
  • PCR product can remain in tube for later analysis
  • Hinged caps provide snug seal
  • Presterilized; RNase- and DNase-free
Recommended for: Complex DNA or cDNA, very low copy number targets, more than 30 thermal cycles, multiple PCR or RT-PCR
Specifications
Specifications
Thermal Cyclers
Sterile