missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Howard Leight™ Leightning™ L3 Headband Earmuffs
Click to view available options
Leightning L3 Earmuffs
Description
- Exclusive padded foam headband and super soft ear cushions eliminate pressure on head
- Air Flow Control™ technology delivers optimal attenuation across all frequencies
- Robust steel-wire construction withstands demanding use
- Snap-in ear cushions replace easily
- Telescoping height adjustment offers personalized comfort

Specifications
Specifications
Color | Black |
Includes | Padded foam headband and super soft ear cushions |
For Use With (Application) | Agriculture and Farming, Building Construction, Forestry, General Contracting, Heavy Construction, Landscaping, Lumber or Wood Products, Metal Fabrication, Petrochemical Steel |
Noise Reduction Rating (NRR) | 34 dB |
Product Type | Ear Muffs |
Material | Metal |
Style | Over-the-head |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Honeywell™ Howard Leight™ Leightning™ L3 Headband Earmuffs >
Spot an opportunity for improvement?Share a Content Correction