missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Howard Leight™ Thunder™ T3 Dielectric Ear Muff

Product Code. 12685013
Click to view available options
Thunder T3 Earmuff
This item is not returnable. View return policy
This item is not returnable. View return policy

NRR: 30dB; Class A

  • Suitable for virtually all workplaces, especially electrical and mining environments
  • Air Flow Control™ technology delivers optimal attenuation across all frequencies
  • ABS plastic headband does not conduct electricity
  • Unique dual-headband design with inner-ventilation minimizes pressure on head and allows contact surface to breathe easier in warm/humid climates
  • Nondeforming outer headband withstands toughest environments
  • Snap-in ear cushions make replacement quick and easy

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Certifications/Compliance ANSI S3.19-1974, NRR 30
For Use With (Application) Superior electrical hearing protection, and also snap-in ear cushions for easy maintenance
Noise Reduction Rating (NRR) 30 dB
Product Type Thunder T3 Ear Muff
Dielectric Yes
Material ABS Plastic
Style Over-the-head
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'