missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA7 Mouse anti-Human, CoraLite 488, Clone: 2A7H6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67112-20UL
This item is not returnable.
View return policy
Description
HOXA7 Monoclonal antibody apecifically detetects HOXA7 antigen in Human Samples. It is validated for Immunofluorescence.Specifications
HOXA7 | |
Monoclonal | |
1000 ug/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
HOXA7 | |
HOXA7 fusion protein Ag19025 | |
20 μL | |
3204 | |
Store at -20°C. Avoid exposure to light. | |
IgG1 |
Immunofluorescence | |
2A7H6 | |
CoraLite 488 | |
BC148692 | |
Mouse | |
Protein G purification | |
Primary | |
Human | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 20UL CoraLite488-conjugated HOXA7 Mouse McAb
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1A
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one