missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA7 Mouse anti-Human, Unconjugated, Clone: 2A7H6, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | HOXA7 |
---|---|
Clone | 2A7H6 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849555
|
Proteintech
67112-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16839555
|
Proteintech
67112-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HOXA7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HOXA7. This antibody recognizes human antigen. The HOXA7 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
HOXA7 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
3204 | |
Primary |
2A7H6 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Human | |
IgG1 |