missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540364
This item is not returnable.
View return policy
Description
Peptide sequence: ALSGADEQPP FHPEPRKSDC AQDKSVFGET EEQKCSTPVY PWMQRMNSCN Sequence homology: Dog: 79%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%; Rat: 93%
HOXB6 Polyclonal antibody specifically detects HOXB6 in Human samples. It is validated for Western Blot.Specifications
HOXB6 | |
Polyclonal | |
Unconjugated | |
HOXB6 | |
homeo box 2B; homeo box B6; homeobox B6; homeobox protein Hox-2.2; Homeobox protein Hox-2B; homeobox protein Hox-B6; Homeobox protein Hu-2; homeobox protein MH-22A; HOX2; Hox-2.2; HOX2B; Hoxb6; Hoxb-6; HU-2; RGD1562142 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3216 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P17509 | |
HOXB6 | |
Synthetic peptide directed towards the N-terminal of human HOXB6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction