missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565913
This item is not returnable.
View return policy
Description
Immunogen sequence: SYFTNPSLSCH LAGGQDVLPN VALNSTAYDP VRHFSTYGAA VAQNRIYSTP FYSPQENVVF SSSRGPYDYG SNSFYQEKDM LSNCRQNTLG HNTQTSIAQD FSSEQ Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
This protein belongs to the homeobox family, members of which encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene, HOXC6, is one of several HOXC genes located in a cluster on chromosome 12. Three genes, HOXC5, HOXC4 and HOXC6, share a 5' non-coding exon.Specifications
HOXC6 | |
Polyclonal | |
Unconjugated | |
HOXC6 | |
Antennapedia-type homeotic factor; CP25; HHO.C8; homeo box 3C; homeo box C6; homeo box C8 protein; homeobox C6; homeobox protein CP25; Homeobox protein HHO.C8; homeobox protein Hox-3.3; Homeobox protein Hox-3C; homeobox protein Hox-6.1; Homeobox protein Hox-C6; Hox C6; HOX3; Hox-3.3; HOX3C; Hox-6.1; Hoxc6; Hoxc-6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3223 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09630 | |
HOXC6 | |
Recombinant Human HOXC6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only