missing translation for 'onlineSavingsMsg'
Learn More

HP Rotaflo™, Stopcock Spare Parts - Replacement Keys

Product Code. p-8338003
Click to view available options
Plug Bore:
3 mm
6 mm
10 mm
Unit Size:
Each
Pack of 2
This item is not returnable. View return policy

Product Code. 10719555

Brand: Quickfit HP3K

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Replacement keys for stopcock

Available in 3mm and 6mm bore sizes
TRUSTED_SUSTAINABILITY

Specifications

Type Stopcock Key
Plug Bore 3 mm
Material Borosilicate Glass
For Use With (Equipment) HP Rotaflo™ stopcock
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.