Learn More
SCAT HPLC Starter Kits - Supply, V2.0
SCAT Safety Caps are the standard for safely supplying your HPLC with solvents. They block harmful vapours and protect your HPLC system from contamination.HPLC starter kit equipped with SafetyCaps, blanking plugs, fittings and vent valves.
Brand:  SCAT 399200
266.69 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
 View more versions of this product
Product Code. 16463819
  • £307.00 / 1 set
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Original brand quality from the premium manufacturer SCAT
  • Initial removal equipment that can be used with all common HPLC system
  • Starter kit consisting of SCAT SafetyCaps, blind plugs, fittings and air valves for the supply of HPLC systems
  • All set components can be reordered separately
starter kit, hplc system, safetycaps, blind plugs, fittings, air valve
Specifications
Specifications
Supply of HPLC Systems
3 x SafetyCaps I (307019), 1 x SafetyCap II (307909), 1 x blind plug, 5 x fittings 1.6/2.3/3.2 mm, 4 x air valves (317010)
HPLC Starter Kit 1
Any HPLC System
GL45
Product Suggestions
Product Suggestions
Videos
Videos
  • HOW TO Install Safety Caps on your HPLC
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SCAT HPLC Starter Kits - Supply, V2.0 > HPLC Starter Kit 1 - Supply, V2.0