missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT HPLC Starter Kits - Supply, V2.0
Shop All SCAT Europe Products
Click to view available options
Type:
HPLC Starter Kit 1
HPLC Starter Kit 2
HPLC Starter Kit 3
Solvent Waste Kit
Unit Size:
1 set
Pack of 4
Description
- Original brand quality from the premium manufacturer SCAT
- Initial removal equipment that can be used with all common HPLC system
- Starter kit consisting of SCAT SafetyCaps, blind plugs, fittings and air valves for the supply of HPLC systems
- All set components can be reordered separately
starter kit, hplc system, safetycaps, blind plugs, fittings, air valve
Specifications
Specifications
| For Use With (Equipment) | Any HPLC System |
| Includes | 4 x SafetyCaps II (307909), 4 x blind plugs, 8 x fittings 1.6/2.3/3.2 mm, 4 x air valves (317010) |
| Thread Size | GL45 |
| For Use With (Application) | Supply of HPLC Systems |
| Type | HPLC Starter Kit 2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction