missing translation for 'onlineSavingsMsg'
Learn More

SCAT HPLC Starter Kits - Supply, V2.0

Product Code. 16687493 Shop All SCAT Europe Products
Click to view available options
Type:
HPLC Starter Kit 1
HPLC Starter Kit 2
HPLC Starter Kit 3
Solvent Waste Kit
Unit Size:
1 set
Pack of 4

Product Code. 16687493

Brand: SCAT 399202

Only null left
Add to basket
Add to basket

SCAT Safety Caps are the standard for safely supplying your HPLC with solvents. They block harmful vapours and protect your HPLC system from contamination.HPLC starter kit equipped with SafetyCaps, blanking plugs, fittings and vent valves.

  • Original brand quality from the premium manufacturer SCAT
  • Initial removal equipment that can be used with all common HPLC system
  • Starter kit consisting of SCAT SafetyCaps, blind plugs, fittings and air valves for the supply of HPLC systems
  • All set components can be reordered separately

starter kit, hplc system, safetycaps, blind plugs, fittings, air valve

Specifications

For Use With (Equipment) Any HPLC System
Includes 4 x SafetyCaps III (307910), 8 x blind plugs, 12 x fittings 1.6/2.3/3.2 mm, 4 x air valves (317010)
Thread Size GL45
For Use With (Application) Supply of HPLC Systems
Type HPLC Starter Kit 3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.