Learn More
SCAT HPLC Starter Kits - Supply, V2.0
SCAT Safety Caps are the standard for safely supplying your HPLC with solvents. They block harmful vapours and protect your HPLC system from contamination.HPLC starter kit equipped with SafetyCaps, blanking plugs, fittings and vent valves.
Brand: SCAT 399203
This item is not returnable.
View return policy
Description
- Original brand quality from the premium manufacturer SCAT
- Initial removal equipment that can be used with all common HPLC system
- Starter kit consisting of SCAT SafetyCaps, blind plugs, fittings and air valves for the supply of HPLC systems
- All set components can be reordered separately
starter kit, hplc system, safetycaps, blind plugs, fittings, air valve
Specifications
HPLC Starter Kit 4: 4 x Safety Cap IV, GL45, 4 x air valve, 4 x change indicator, 16 x PFA fitting 1.6 mm OD, green, 16 x PFA fitting 2.3 mm OD, violet, 16 x PFA fitting 3.2 mm OD, blue, 12 x PFA blind plugs, 4 x labelling field | |
4 Pk. | |
Solvent Waste Kit |
PTFE | |
GL45 |
Product Content Correction
Product Title
SCAT HPLC Starter Kits - Supply, V2.0 > Includes: HPLC Starter Kit 4
Spot an opportunity for improvement?Share a Content Correction