Learn More
SCAT HPLC Starter Kits - Supply, V2.0
Product Code. 18696458 Shop All SCAT Europe Products
Click to view available options
Type:
HPLC Starter Kit 1
HPLC Starter Kit 2
HPLC Starter Kit 3
Solvent Waste Kit
Unit Size:
1 set
Pack of 4
Product Code. 18696458
Brand: SCAT 399204
Only null left
Add to basket
Add to basket
SCAT Safety Caps are the standard for safely supplying your HPLC with solvents. They block harmful vapours and protect your HPLC system from contamination.HPLC starter kit equipped with SafetyCaps, blanking plugs, fittings and vent valves.
  • Original brand quality from the premium manufacturer SCAT
  • Initial removal equipment that can be used with all common HPLC system
  • Starter kit consisting of SCAT SafetyCaps, blind plugs, fittings and air valves for the supply of HPLC systems
  • All set components can be reordered separately
starter kit, hplc system, safetycaps, blind plugs, fittings, air valve
Specifications
Includes HPLC Starter Kit 6: 4 x Safety Cap VI, GL45, 4 x air valve, 4 x change indicator, 24 x PFA fitting 1.6 mm OD, green, 24 x PFA fitting 2.3 mm OD, violet, 24 x PFA fitting 3.2 mm OD, blue, 20 x PFA blind plugs, 4 x labelling field
Material PTFE
Thread Size GL45
Type Solvent Waste Kit
Quantity 4 Pk.