Learn More
SCAT HPLC Supply-Sets, V2.0
The HPLC Supply Sets, V2.0 of the manufacturer SCAT are combinations of equipment for contamination-free supply of HPLC systems with solvents. The sets consist of a laboratory bottle with GL 45 thread, a SCAT SafetyCap as well as capillaries, suction filters and air valves.
£124.00 - £218.00
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
16413799
|
SCAT
307312 |
Set I | 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle round & brown glass + 1 x 1.5 m capillary 3.2 mm OD, 1 x PTFE suction filter + air valve (317010) |
£171.00
1 set |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
16433789
|
SCAT
307300 |
Set I | 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle round & clear glass + 1 x 1.5 m capillary 3.2 mm OD, 1 x PTFE suction filter + air valve (317010) |
£125.00
1 set |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
16443789
|
SCAT
307301 |
Set I | 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle square & clear glass + 1 x 1.5 m capillary 3.2 mm OD, 1 x PTFE suction filter + air valve (317010) |
£124.00
1 set |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
16423799
|
SCAT
307313 |
Set II | 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle round & brown glass + 2 x 1.5 m capillaries 3.2 mm OD, 2 x PTFE suction filters + air valve (317010) |
£200.00
1 set |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
16453789
|
SCAT
307303 |
Set II | 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle round & clear glass + 2 x 1.5 m capillaries 3.2 mm OD, 2 x PTFE suction filters + air valve (317010) |
£141.00
1 set |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
16473789
|
SCAT
307305 |
Set II | 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle square & clear glass + 2 x 1.5 m capillaries 3.2 mm OD, 2 x PTFE suction filters + air valve (317010) |
£147.00
1 set |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
16433799
|
SCAT
307314 |
Set III | 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle round & brown glass + 3 x 1.5 m capillaries 3.2 mm OD, 3 x PTFE suction filters + air valve (317010) |
£218.00
1 set |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
16463789
|
SCAT
307304 |
Set III | 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle round & clear glass + 3 x 1.5 m capillaries 3.2 mm OD, 3 x PTFE suction filters + air valve (317010) |
£165.00
1 set |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
16483789
|
SCAT
307306 |
Set III | 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle square & clear glass + 3 x 1.5 m capillaries 3.2 mm OD, 3 x PTFE suction filters + air valve (317010) |
£171.00
1 set |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
16453799
|
SCAT
307347 |
Set III | 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle Youtility & clear glass + 3 x 1.5 m capillaries 3.2 mm OD, 3 x PTFE suction filters + air valve (317010) |
£171.00
1 set |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17927260
|
SCAT™
307327 |
Complete Solvent Collection System | 1 x SafetyCap II GL28, 1x 100 mL Laboratory bottle round, brown glass, 2 x 1.5 m capillaries 3.2 mm OD, 2 x PTFE suction filters, air valve (317010) |
£141.10
1 set |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Customised system with high quality set components
- SafetyCaps of the premium manufacturer SCAT
- Laboratory bottles
- Price attractive in comparison with single purchase
- Ready for use, no further components necessary
SCAT offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.
Specifications
Any HPLC System | |
GL45 |