Learn More
SCAT™ HPLC Supply and Waste Set, V2.0, Suitable for All HPLC Systems
The all-round carefree package suitable for all HPLC and UHPLC systems.
Brand:  SCAT™ 307447
536.05 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 5.28000kg
 View more versions of this product
Product Code. 17865704
  • £710.00 / 1 set
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • Protects your system from contamination
  • Keeps solvent mixtures stable
  • High performance materials with excellent chemical resistance
The safety set connects tubing safely and blocks hazardous vapours. Air valves and exhaust filters provide safe pressure equalisation without under- or overpressure in solvent bottles and waste containers
Specifications
Specifications
4 x Safety Cap III, GL45, 4 x air valve, 1 x Safety Waste Cap, S60/61, electrostatic conductive, 1 x Exhaust filter M, V3.0, with splash guard and change indicator, 1 x 5 liter canister, PE-HD-EL, 1 x Grounding cable, 2 x clamp, 1.5 m length, 4 x Laboratory bottle DURAN, GL45, 1000 mL, clear glass and round, with cap and pouring ring, 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 to 8 mm ID, 1 x connector, curved, 5 to 11.5 mm ID, 1 x connector, angled, 9.5 to 10 mm ID
HPLC System
Any HPLC System