missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ HPLC Waste Sets, V2.0

Product Code. p-7214837 Shop All SCAT Europe Products
Click to view available options
Capacity (Metric):
2.50 L
5 L
10 L
12 L
20 L
30 L
Material:
HDPE
HDPE-EC
Polypropylene
UN-X HDPE
Thread Size:
GL45
S50
S51
S55
S60/61
S70/71
S90
No. of Ports:
3 (2 Capillary, 1 Tubing)
5 (4 Capillary, 1 Tubing)
6 (3 Capillary, 3 Tubing)
Unit Size:
1 set
Each

Product Code. 16493789

Brand: SCAT™ 307307

Only null left
Add to basket
Add to basket

HPLC Waste Sets, V2.0 are economical combinations for clean and safe disposal of liquid laboratory waste by HPLC systems or other analytical instruments. All sets consist of a canister and an original SCAT™ SafetyCap with exhaust filter as well as fittings, blind plugs and a tube connector.

SCAT™ offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.

  • Customized system with high-quality set components
  • Safety canisters made of HDPE or polypropylene material
  • Attractive price compared to single purchase
  • Ready to use, no further components necessary

Specifications

For Use With (Equipment) HPLC Systems
Type Complete Solvent Collection System
Solvent Channels 3
Material HDPE
Includes Safety Waste Cap, 2 x 2.3/3.2 mm OD, 1 x 6.4 to 9 mm ID, HDPE Canister, Exhaust Filter M with Splash Guard and Exchange Indicator
Color White
Thread Size GL45
Capacity (Metric) 5 L
No. of Ports 3 (2 Capillary, 1 Tubing)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.