missing translation for 'onlineSavingsMsg'
Learn More

HPR Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

£121.00 - £352.00

Specifications

Antigen HPR
Concentration 0.5 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17793294
View Documents
Invitrogen
PA5121901
20 μL
£121.00
20µL
Please to purchase this item. Need a web account? Register with us today!
17783294
View Documents
Invitrogen
PA5121900
100 μL
£352.00
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a haptoglobin-related protein that binds hemoglobin as efficiently as haptoglobin. Unlike haptoglobin, plasma concentration of this protein is unaffected in patients with sickle cell anemia and extensive intravascular hemolysis, suggesting a difference in binding between haptoglobin-hemoglobin and haptoglobin-related protein-hemoglobin complexes to CD163, the hemoglobin scavenger receptor. This protein may also be a clinically important predictor of recurrence of breast cancer.
Specifications

Specifications

HPR
Western Blot
Unconjugated
Rabbit
Human, Rat
P00739
3250
Gln102-Glu347 (Accession P00739), with N-terminal His Tag
Primary
-20°C
HPR
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300
A-259H10.2; Haptoglobin-related locus; haptoglobin-related protein; HP; HPR
HPR
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications