Learn More
Applied Biosystems™ HRM Software Module for QuantStudio™ 12K Flex System
This software module expands the functionality of the QuantStudio™ 12K Flex software, allowing high resolution melting (HRM) analysis to be performed on PCR amplicons.
Brand: Applied Biosystems™ 4472054
Description
This software module expands the functionality of the QuantStudio™ 12K Flex software, allowing high resolution melting (HRM) analysis to be performed on PCR amplicons.
High Resolution Melt (HRM) analysis is a new, post-PCR analysis method used for identifying genetic variation in nucleic acid sequences. Simple and fast, this method is based on PCR melt (dissociation) curve techniques and is enabled by the recent availability of improved double-stranded DNA (dsDNA)-binding dyes along with next-generation real-time PCR instrumentation and analysis software. HRM analysis can discriminate DNA sequences based on their composition, length, GC content, or strand complementarity.
Specifications
QuantStudio™ 12 k Flex | |
QuantStudio™ | |
Windows 7 | |
10 Seat |
For Research Use Only. Not for use in diagnostic procedures.