missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hs6st3 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547733
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
Hs6st3 Polyclonal antibody specifically detects Hs6st3 in Mouse samples. It is validated for Immunohistochemistry (Frozen).Specifications
Hs6st3 | |
Polyclonal | |
Unconjugated | |
Hs6st3 | |
6OST3; 6-ost-3; DKFZp761K2315; heparan sulfate 6-O-sulfotransferase 3; heparan sulfate 6-O-sulfotransferase 3b; heparan-sulfate 6-O-sulfotransferase 3; heparan-sulfate 6-O-sulfotransferase 3 pseudogene; heparan-sulfate 6-O-sulfotransferase 3-B; HS 6-OST-3-B; hs6st3; HS6ST-3; hs6st3b; RGD1560050; sb:eu962 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
50787 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen) | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9QYK4 | |
Hs6st3 | |
CHO-derived recombinant mouse Heparan Sulfate 6-O-Sulfotransferase/HS6ST3 Pro28-Trp470 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |