Learn More
HSBP1L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561698
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NLFQELQEHF QALTATLNLR MEEMGNRIED LQKNVNDLMV QAGIENSIKE QMLE Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 81%.
HSBP1L1 Polyclonal antibody specifically detects HSBP1L1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
HSBP1L1 | |
Polyclonal | |
Unconjugated | |
HSBP1L1 | |
heat shock factor binding protein 1 like 1; heat shock factor binding protein 1-like 1; heat shock factor-binding protein 1-like protein 1; HSBP1L1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
440498 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
C9JCN9 | |
HSBP1L1 | |
Recombinant protein corresponding to Human HSBP1L1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only