Learn More
HSD11B2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559904
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GDMPYPCLGA YGTSKAAVAL LMDTFSCELL PWGVKVSIIQ PGCFKTESVR NVGQWEKRKQ LLLANLPQEL LQAYGKDYIE HLHGQFLHSL R Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 89%.
There are at least two isozymes of the corticosteroid 11-beta-dehydrogenase, a microsomal enzyme complex responsible for the interconversion of cortisol and cortisone. The type I isozyme has both 11-beta-dehydrogenase (cortisol to cortisone) and 11-oxoreductase (cortisone to cortisol) activities. The type II isozyme, encoded by this gene, has only 11-beta-dehydrogenase activity. In aldosterone-selective epithelial tissues such as the kidney, the type II isozyme catalyzes the glucocorticoid cortisol to the inactive metabolite cortisone, thus preventing illicit activation of the mineralocorticoid receptor. In tissues that do not express the mineralocorticoid receptor, such as the placenta and testis, it protects cells from the growth-inhibiting and/or pro-apoptotic effects of cortisol, particularly during embryonic development. Mutations in this gene cause the syndrome of apparent mineralocorticoid excess and hypertension.Specifications
HSD11B2 | |
Polyclonal | |
Unconjugated | |
HSD11B2 | |
11(beta)-HSD2; 11-beta-HSD; 11-beta-HSD type II; 11-beta-HSD2; 11-beta-hydroxysteroid dehydrogenase type 2; 11-beta-hydroxysteroid dehydrogenase type II; 11-DH2; 11-HSD type II; 11HSD2; AME; AME1; corticosteroid 11-beta-dehydrogenase isozyme 2; -HSD11 type II; HSD11B2; HSD11K; HSD2; hydroxysteroid (11-beta) dehydrogenase 2; hydroxysteroid 11-beta dehydrogenase 2; Hydroxysteroid dehydrogenase, 11 beta type 2; NAD-dependent 11-beta-hydroxysteroid dehydrogenase; SDR9C3; Short chain dehydrogenase/reductase family 9C member 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3291 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P80365 | |
HSD11B2 | |
Recombinant protein corresponding to Human HSD11B2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only