missing translation for 'onlineSavingsMsg'
Learn More

HSDL2 Monoclonal Antibody (2A11BG6AH6), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen A21977

Code : 50

Additional Details : Weight : 0.01000kg

Product Code. 11390742

  • £394.00 / Each
Estimated Shipment: 12-08-2024
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

HSDL2 Monoclonal antibody specifically detects HSDL2 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation.
Specifications

Specifications

HSDL2
Monoclonal
1 mg/mL
HEPES buffered saline with 0.02% sodium azide
Q6YN16
HSDL2
Human HSDL2
100 μg
Primary
Human
Antibody
IgG1
ELISA, Immunocytochemistry, Immunoprecipitation
2A11BG6AH6
Unconjugated
HSDL2
2610207I16Rik; C9orf99; Hsdl2; hydroxysteroid dehydrogenase like 2; hydroxysteroid dehydrogenase-like protein 2; RGD1305387; SDR13C1; short chain dehydrogenase/reductase family 13C member 1; short chain dehydrogenase/reductase family 13C, member 1
Mouse
purified
RUO
84263
4° C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers