Learn More
HSF4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565521
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: GKGSFSPEGPR NAQQPEPGDP REIPDRGPLG LESGDRSPES LLPPMLLQPP QESVEPAGPL DVLGPSLQGR EWTLMDLDME LSLMQPLVPE RGEPELAVKG LNSPSP Highest antigen sequence identity to the following orthologs - mouse 74%, rat 74%.
Heat-shock transcription factors (HSFs) activate heat-shock response genes under conditions of heat or other stresses. HSF4 lacks the carboxyl-terminal hydrophobic repeat which is shared among all vertebrate HSFs and has been suggested to be involved in the negative regulation of DNA binding activity. Two alternatively spliced transcripts encoding distinct isoforms and possessing different transcriptional activity have been described.Specifications
HSF4 | |
Polyclonal | |
Unconjugated | |
HSF4 | |
ATHSF4; AT-HSFB1; binds heat shock DNA elements; cataract; CTM; CTRCT5; DNA binding protein; heat shock factor 4; heat shock factor protein 4; Heat shock transcription factor 4; hHSF4; HSF 4; Hsf4; HSFB1; HSTF 4; HSTF4; ldis1; Marner; mHSF4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3299 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULV5 | |
HSF4 | |
Recombinant Human HSF4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only