Learn More
HSP20 Mouse anti-Human, Unconjugated, Clone: 1G7D2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67327-1-IG-150UL
This item is not returnable.
View return policy
Description
The HSP20 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HSP20. This antibody recognizes human, mouse, rat, pig antigen. The HSP20 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
HSP20 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, ELISA | |
1G7D2 | |
Unconjugated | |
150 μL | |
126393 | |
IgG2a |