Learn More
HSP40 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527604
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (93%), Rat (94%), Pig (91%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.Specifications
HSP40 | |
Polyclonal | |
Unconjugated | |
DNAJB1 | |
0610007I11Rik; DjB1; DnaJ (Hsp40) homolog, subfamily B, member 1; DnaJ heat shock protein family (Hsp40) member B1; dnaJ homolog subfamily B member 1; DnaJ protein homolog 1; DNAJ1; Dnajb1; HDJ1; hDj-1; Heat shock 40 kDa protein 1; Heat shock protein; HSP; HSP 40; HSP40; HSPF1; human DnaJ protein 1; NDAJB1; radial spoke 16 homolog B; RSPH16B; Sis1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3337 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.69 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P25685 | |
DNAJB1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 194 of Human Hsp40 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |