Learn More
HSPA1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA529605
Code : PN
Detalles adicionales : Weight : 0.01000kg
Descripción
Recommended positive controls: A431, HeLa, U87-MG, SK-N-SH, IMR32, SK-N-AS. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This intronless gene encodes a 70 kDa heat shock protein which is a member of the heat shock protein 70 family. In conjunction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.Especificaciones
HSPA1B | |
Polyclonal | |
Unconjugated | |
Hspa1b | |
68 kDa heat shock protein; Hcp70.1; Heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; Heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; heat shock 70kD protein 1B; heat shock 70kD protein 1B (mapped); heat shock 70kDa protein 1B; Heat shock protein; heat shock protein 1B; heat shock protein 70.1; heat shock protein family A (Hsp70) member 1B; heat shock protein, 70 kDa 1; HSP; hsp68; Hsp70; HSP70.1; HSP70.1/HSP70.2; HSP70.2; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; HSP70-1B; HSP70-2; Hsp70a1; HSP70I; Hsp72; HSPA1; Hspa1a; HSPA1B; Hspa2; HSX70 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
294254, 3304 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal, 0.025% ProClin 300; pH 7 | |
P0DMV9, P0DMW1 | |
Hspa1b | |
Recombinant protein fragment corresponding to a region within amino acids 190 and 423 of Human HSP70 1B | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |