Learn More
HSPB7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555670
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SPEDIIVTTS NNHIEVRAEK LAADGTVMNT FAHKCQLPED VDPTSVTSAL REDGSLTIRA RRHPHTEHVQ QTFRTEIK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
HSPB7 encodes a small heat shock family B member that can heterodimerize with similar heat shock proteins. Defects in this gene are associated with advanced heart failure. In addition, the encoded protein may be a tumor suppressor in the p53 pathway, with defects in this gene being associated with renal cell carcinoma.Specifications
HSPB7 | |
Polyclonal | |
Unconjugated | |
HSPB7 | |
27kDa; Cardiovascular heat shock protein; cvHsp; heat shock 27kD protein family member 7; heat shock 27kD protein family, member 7 (cardiovascular); heat shock 27kDa protein family member 7 (cardiovascular); heat shock 27kDa protein family, member 7 (cardiovascular); Heat shock protein; Heat shock protein 25 kDa 2; heat shock protein 25 kDa 2 (cardiovascular); heat shock protein B7; heat shock protein beta-7; heat shock protein family B (small) member 7; heat shock protein family, alpha-crystallin-related, b7; heat shock protein family, member 7 (cardiovascular); HSP; Hsp25-2; HspB7; Protein p19/6.8; zgc:103426 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27129 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UBY9 | |
HSPB7 | |
Recombinant protein corresponding to Human HSPB7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only