Learn More
Julabo™ HST Booster Heater for Ultra-Low Refrigerated Circulators
Brand:  Julabo™ 8810011
Code : A0
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11984798
  • £3615.00 / Each
Estimated Shipment: 14-08-2024
to see stock.
Add to basket
Specifications
FP51-SL Ultra-Low Refrigerated-Heating CirculatorI
HST Booster Heater
Ideal for controlling the temperature of large or distant objects