missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup
HT ExoSAP-IT Fast High-Throughput PCR Product Clean-up, with its shortened 14 minute protocol, is a highly stable, customized formulation of the original ExoSAP-IT reagent
£710.00 - £2985.00
Specifications
Content And Storage | -20°C |
---|---|
Format | 8 Tube Strip, 96-well Plate, Vial |
Final Product Type | Fast High-Throughput PCR Product Clean-up |
For Use With (Application) | For high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes |
Product Type | HT ExoSAP-IT™ Fast High-Throughput PCR Product Clean-up |
Product Code | Brand | Description | For Use With (Application) | No. of Reactions | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Application) | No. of Reactions | Price | Quantity & Availability | |||
15869916
|
Applied Biosystems™
785955000RXN |
Fast High-Throughput PCR Product Clean-up, 5000 rxns, Highly Stable, 4 Minute Protocol, Ideal for High-throughput PCR Product Clean-up Using Liquid Handling Platforms and Multi-Channel Pipettes | For high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes | 5000 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15829916
|
Applied Biosystems™
785951000RXN |
Fast High-Throughput PCR Product Clean-up, 1000 rxns, Highly Stable, 4 Minute Protocol, Ideal for High-throughput PCR Product Clean-up Using Liquid Handling Platforms and Multi-Channel Pipettes | For high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes | 1000 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- 14 minute protocol is half the time of standard protocols
- One-tube PCR clean-up - add HT ExoSAP-IT Fast reagent directly to PCR product
- 100% sample recovery - no loss of PCR products regardless of the fragment size
- Stable at 4°C for one month
- Removes excess primers and dNTPs - does not interfere with downstream applications sequencing, SNP analysis, single base extension, fragment analysis
- It is ideal for high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes
- Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
Specifications
-20°C | |
Fast High-Throughput PCR Product Clean-up | |
HT ExoSAP-IT™ Fast High-Throughput PCR Product Clean-up | |
14 min |
8 Tube Strip, 96-well Plate, Vial | |
For high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes | |
Each |
For Research Use Only. Not for use in diagnostic procedures.