Learn More
HTBA, MedChemExpress
MedChemExpress HTBA(3-Hydroxy-2,4,6-triiodobenzoic acid) for your research needs.
Brand:  Medchem Express HY-15919
Encompass_Preferred
 View more versions of this product
Product Code. 16436996
  • £68.97 / 100mg
Explore more special offers
Description
Description
HTBA(3-Hydroxy-2,4,6-triiodobenzoic acid) for your research needs.
Specifications
Specifications
HTBA
Off-White
C7H3I3O3
DMSO : ≥ 33 mg/mL (63.98 mM)
515.81
99.61%
Solid
53279-72-4
Research
100 mg
O=C(O)C1=C(I)C=C(I)C(O)=C1I
515.81
Research