missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR7 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519124
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
5-HT7 Receptor is a Serotonin Receptor with several functions, including regulation of the circadian rhythm and mediation of smooth-muscle relaxation in the vasculature and GI tract. This receptor has been implicated in depression, autism, and other neuropsychiatric disorders. Three protein isoforms have been documented: isoform a: 445aa; isoform b: 432aa; isoform d: 479aa. 5-HT7 Receptor has been reported in aorta, brain, heart, placenta, small intestine, spleen, artery, and vessel. An EST has been isolated from a testis library.Specifications
HTR7 | |
Polyclonal | |
Unconjugated | |
HTR7 | |
5-HT receptor 7a; 5-HT receptor 7b; 5Ht7; 5-HT7; 5-HT-7; 5HT7 Receptor; 5-HT7 receptor; 5-HT-X; 5-hydroxytryptamine (serotonin) receptor 7; 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled); 5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled; 5-hydroxytryptamine receptor 7; GPRFO; high affinity serotonin receptor (5HT7); HT7; Htr7; OTTHUMP00000020077; OTTHUMP00000020078; OTTHUMP00000020079; Serotonin receptor 7 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
3363 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P34969 | |
HTR7 | |
Synthetic peptide sequence (QNADYCRKKGHDS) corresponding to the C-terminus amino acids of HTR7 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |