missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTRA2 Monoclonal Antibody (196C429), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA141040
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: HL-60 or Jurkat whole cell lysate.
The inhibitor of apoptosis (IAP) family of proteins regulates programmed cell death triggered by various stimuli. All IAPs have at least one baculovirus IAP repeat (BIR) motif that is essential for their anti-apoptotic activity (1-3). Recently, a serine protease has been isolated, which is released from the mitochondria upon induction of apoptosis by apoptotic stimuli. This protein called Omi/HtrA2 is a mitochondrial protein that binds to IAP (4). Like Smac/DIABLO, the mature Omi protein contains a conserved IAP-binding motif (AVPS) at its N terminus. The deregulated expression of Omi in the cytoplasm of mammalian cells induces apoptosis indicating that Omi could participate in the mitochondrial apoptotic pathway.Specifications
HTRA2 | |
Monoclonal | |
Unconjugated | |
HTRA2 | |
AI481710; High temperature requirement protein A2; HtrA serine peptidase 2; HtrA2; HtrA-like serine protease; mnd2; motor neuron degeneration 2; OMI; Omi stress-regulated endoprotease; PARK13; protease, serine, 25; Prss25; Serine protease 25; serine protease HTRA2, mitochondrial; serine protease OMI; serine proteinase OMI | |
Mouse | |
Protein G | |
RUO | |
27429 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
196C429 | |
PBS with 0.05% BSA and 0.05% sodium azide | |
O43464 | |
HTRA2 | |
Synthetic peptide corresponding to residues 335-350 of human Htra2. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |