missing translation for 'onlineSavingsMsg'
Learn More

HuB/HuD Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA538596

Code : NEW

Additional Details : Weight : 0.00900kg

Product Code. 15595511

  • £402.00 / 100µg
Estimated Shipment: 02-09-2024
to see stock.

Description

Description

Anti-Hu antibodies were originally isolated from a patient with paraneoplastic encephalomyelitis, a rare condition of selective neural tissue injury associated with small cell lung carcinoma. The antibodies bind specifically to antigens present exclusively in neuronal cells and are thus useful as markers of neuronal cells in tissue. The Hu antigen is an RNA-binding protein of the embryonic lethal abnormal visual (Elav) family.
Specifications

Specifications

HuB/HuD
Polyclonal
Unconjugated
ELAVL2
ELAV-like protein 3, Hu-antigen C, HuC, Paraneoplastic cerebellar degeneration-associated antigen, Paraneoplastic limbic encephalitis antigen 21, ELAVL3, HUC, PLE21, ELAV-like protein 4, Hu-antigen D, HuD, Paraneoplastic encephalomyelitis antigen HuD, ELAVL4, HUD, PNEM
Rabbit
Antigen affinity chromatography
RUO
15569, 15572, 1993, 1996, 286973, 432358
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
P26378, Q12926, Q60899, Q61701, O09032, Q8CH84
ELAVL2, ELAVL4
A synthetic peptide derived from the internal region of human ELAV2/4
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers